Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: UGT2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105677.2 | NP_001099147.2 | 6 | 1355 | ||
NM_001301233.1 | NP_001288162.1 | 5 | 1223 | |||
GenBank mRNA | FJ664272.1 | - | 6 | 1355 | ||
FJ664273.1 | - | 5 | 1223 |
Gene Symbol: UGT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252274.2 | NP_001239203.2 | 7 | 1940 | ||
NM_001252275.2 | NP_001239204.2 | 7 | 1442 | |||
NM_001301239.1 | NP_001288168.1 | 5 | 1248 | |||
NM_006798.4 | NP_006789.3 | 7 | 1442 | |||
GenBank mRNA | AJ006054.1 | - | 6 | 1389 | ||
AK304249.1 | - | 7 | 1940 | |||
AK314209.1 | - | 7 | 1440 | |||
BC142640.1 | - | 5 | 1310 | |||
CD105882.1 | - | 3 | 327 |