Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2B SIRNA Assays ›
Gene Symbol:
RABL2B
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2B
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
-
Chromosome Location:
Chr.22: 50767492 - 50783705 on Build GRCh38
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.584862
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s22011
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003789.1
NP_001003789.1
9
1843
NM_001130919.1
NP_001124391.1
9
1839
NM_001130920.1
NP_001124392.1
10
1998
NM_001130921.1
NP_001124393.1
10
2091
NM_001130922.1
NP_001124394.1
9
1840
NM_001130923.1
NP_001124395.1
10
2025
NM_007081.2
NP_009012.1
9
1836
XM_005261899.2
XP_005261956.1
10
2122
XM_011530668.1
XP_011528970.1
10
2125
XM_011530669.2
XP_011528971.1
9
1872
XM_011530670.1
XP_011528972.1
9
1889
XM_011530671.1
XP_011528973.1
10
1999
XM_011530673.1
XP_011528975.1
10
2113
XM_011530675.1
XP_011528977.1
10
2092
XM_017028543.1
XP_016884032.1
10
2315
XM_017028544.1
XP_016884033.1
10
2220
XM_017028545.1
XP_016884034.1
10
2161
XM_017028546.1
XP_016884035.1
10
2163
XM_017028547.1
XP_016884036.1
10
2312
XM_017028548.1
XP_016884037.1
9
1869
XM_017028549.1
XP_016884038.1
10
2158
XM_017028550.1
XP_016884039.1
10
1987
XM_017028551.1
XP_016884040.1
8
2031
XM_017028552.1
XP_016884041.1
10
2285
XM_017028553.1
XP_016884042.1
10
2187
XM_017028554.1
XP_016884043.1
10
2131
XM_017028555.1
XP_016884044.1
10
2133
XM_017028556.1
XP_016884045.1
10
2282
XM_017028557.1
XP_016884046.1
10
2128
XM_017028558.1
XP_016884047.1
10
1966
GenBank mRNA
AF095352.1
-
9
1783
AK094341.1
-
3
2165
BC014879.2
-
9
1804
3906749
BC024281.1
-
10
2041
4580599
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306158.1
NP_001293087.1
9
1845
NM_001306161.1
NP_001293090.1
7
1657
NM_013412.3
NP_038198.1
9
1842
XM_005263569.3
XP_005263626.1
9
1886
XM_005263570.3
XP_005263627.1
9
1853
XM_006712207.3
XP_006712270.1
9
1955
XM_006712208.3
XP_006712271.1
9
1844
XM_006712209.3
XP_006712272.1
9
1998
XM_006712215.3
XP_006712278.1
10
2055
XM_011510509.2
XP_011508811.1
9
1846
XM_017003203.1
XP_016858692.1
8
2595
XM_017003204.1
XP_016858693.1
9
2535
XM_017003206.1
XP_016858695.1
10
1996
XM_017003207.1
XP_016858696.1
9
1909
XM_017003208.1
XP_016858697.1
9
1866
XM_017003209.1
XP_016858698.1
9
1850
XM_017003210.1
XP_016858699.1
8
2583
XM_017003211.1
XP_016858700.1
9
1874
XM_017003212.1
XP_016858701.1
8
2580
XM_017003213.1
XP_016858702.1
8
2565
XM_017003214.1
XP_016858703.1
9
2505
XM_017003216.1
XP_016858705.1
9
1968
XM_017003217.1
XP_016858706.1
9
1852
XM_017003218.1
XP_016858707.1
10
2006
XM_017003219.1
XP_016858708.1
9
1919
XM_017003220.1
XP_016858709.1
8
2562
XM_017003221.1
XP_016858710.1
9
1838
XM_017003222.1
XP_016858711.1
10
1993
XM_017003223.1
XP_016858712.1
6
2403
XM_017003224.1
XP_016858713.1
7
1670
XM_017003225.1
XP_016858714.1
7
1661
XM_017003226.1
XP_016858715.1
6
2373
XM_017003227.1
XP_016858716.1
7
1640
XM_017003228.1
XP_016858717.1
8
1768
XM_017003229.1
XP_016858718.1
9
1862
XM_017003230.1
XP_016858719.1
9
1876
XM_017003232.1
XP_016858721.1
8
1723
GenBank mRNA
AF095351.1
-
9
1776
AK092971.1
-
3
2574
AL137649.1
-
5
1443
BC111008.1
-
7
1647
5769737
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-55874ff697-8pchx:80/10.239.164.120:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0