Search
Search

Arhgef7| Gene Symbol: Arhgef7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113521.1 | NP_001106993.1 | 5 | 641 | ||
| NM_001113522.1 | NP_001106994.1 | 5 | 641 | |||
| NM_053740.2 | NP_446192.2 | 2 | 275 | |||
| XM_008771376.1 | XP_008769598.1 | 5 | 11613 | |||
| XM_008771377.1 | XP_008769599.1 | 5 | 11612 | |||
| XM_008771378.1 | XP_008769600.1 | 5 | 11613 | |||
| XM_008771379.1 | XP_008769601.1 | 698 | ||||
| XM_008771380.1 | XP_008769602.1 | 611 | ||||
| GenBank mRNA | AF044673.1 | - | 2 | 275 | ||
| AY034823.1 | - | 2 | 275 | |||
| AY996221.1 | - | 2 | 275 | |||
| BC131844.1 | - | 3 | 328 | |||