Search
Search

Slc16a10| Gene Symbol: Slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138831.1 | NP_620186.1 | 6 | 1628 | ||
| XM_008772974.1 | XP_008771196.1 | 6 | 1667 | |||
| XM_008772975.1 | XP_008771197.1 | 5 | 1522 | |||
| GenBank mRNA | AB047324.1 | - | 6 | 1628 | ||