Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 6 | 985 | ||
NM_001024960.2 | NP_001020131.1 | 6 | 840 | |||
NM_001198780.1 | NP_001185709.1 | 6 | 839 | |||
NM_005718.4 | NP_005709.1 | 6 | 1351 | |||
GenBank mRNA | AF006087.1 | - | 6 | 792 | ||
AK074205.1 | - | 1 | 1453 | |||
BC012596.2 | - | 5 | 774 | |||
BX419672.2 | - | 6 | 839 |