Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004344.1 | NP_004335.1 | 4 | 410 | ||
GenBank mRNA | AK311940.1 | - | 4 | 405 | ||
BC005334.1 | - | 4 | 410 | 3961448 | ||
BC013873.2 | - | 4 | 408 | 3836808 | ||
BT007256.1 | - | 3 | 363 | |||
X72964.1 | - | 4 | 410 |