Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFAT5 SIRNA Assays ›
Gene Symbol:
NFAT5
Gene Name:
nuclear factor of activated T-cells 5, tonicity-responsive
Gene Aliases:
NF-AT5, NFATL1, NFATZ, OREBP, TONEBP
Chromosome Location:
Chr.16: 69565952 - 69704666 on Build GRCh38
UniGene ID:
Hs.371987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223154
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFAT5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113178.2
NP_001106649.1
13
4290
NM_006599.3
NP_006590.1
12
4239
NM_138713.3
NP_619727.2
13
4293
NM_138714.3
NP_619728.2
14
4355
NM_173214.2
NP_775321.1
13
4301
NM_173215.2
NP_775322.1
13
4301
XM_006721125.3
XP_006721188.1
12
4964
XM_011522817.2
XP_011521119.1
12
4236
XM_011522818.2
XP_011521120.1
15
4428
XM_011522819.2
XP_011521121.1
15
4010
XM_011522820.2
XP_011521122.1
12
3440
XM_011522822.2
XP_011521124.1
11
3388
XM_017022870.1
XP_016878359.1
12
4588
XM_017022871.1
XP_016878360.1
13
4312
XM_017022872.1
XP_016878361.1
12
4169
XM_017022873.1
XP_016878362.1
12
3778
GenBank mRNA
AB020634.1
-
12
4221
AF089824.1
-
13
4243
AF134870.1
-
10
3675
AF163836.1
-
11
3762
AF346509.1
-
14
5241
AJ005683.1
-
13
3917
AJ005684.1
-
13
3917
AJ243298.1
-
13
4060
AJ243299.1
-
13
4052
AK000312.1
-
1
914
BC131509.1
-
13
4194
40083218
BC146765.1
-
12
4221
9007208
U80231.1
-
1
934
Z97016.1
-
1
1883
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
transcription from RNA polymerase II promoter
response to osmotic stress
signal transduction
excretion
positive regulation of transcription from RNA polymerase II promoter
regulation of calcineurin-NFAT signaling cascade
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0