Search
Search

DLX1| Gene Symbol: DLX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038493.1 | NP_001033582.1 | 2 | 1433 | ||
| NM_178120.4 | NP_835221.2 | 3 | 1633 | |||
| GenBank mRNA | BC036189.1 | - | 3 | 1598 | 5311747 | |
| BC053351.1 | - | 2 | 1310 | 5243874 | ||