Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMOX1 SIRNA Assays ›
Gene Symbol:
HMOX1
Gene Name:
heme oxygenase 1
Gene Aliases:
HMOX1D, HO-1, HSP32, bK286B10
Chromosome Location:
Chr.22: 35381067 - 35394214 on Build GRCh38
UniGene ID:
Hs.517581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223854
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002133.2
NP_002124.1
5
1518
GenBank mRNA
BC001491.2
-
5
1478
3504480
M23042.1
-
1
445
X06985.1
-
5
1491
Back To Top
More Information
Panther Classification:
Molecular Function -
oxygenase
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
endothelial cell proliferation
wound healing involved in inflammatory response
negative regulation of leukocyte migration
heme oxidation
cellular iron ion homeostasis
response to oxidative stress
small GTPase mediated signal transduction
excretion
regulation of blood pressure
cell death
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of muscle cell apoptotic process
smooth muscle hyperplasia
cellular response to nutrient
negative regulation of mast cell cytokine production
erythrocyte homeostasis
low-density lipoprotein particle clearance
regulation of transcription from RNA polymerase II promoter in response to iron
cellular response to heat
response to nicotine
intracellular signal transduction
heme catabolic process
response to hydrogen peroxide
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of mast cell degranulation
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of neuron apoptotic process
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
response to estrogen
positive regulation of chemokine biosynthetic process
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of vasodilation
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
protein homooligomerization
iron ion homeostasis
cellular response to arsenic-containing substance
cellular response to cadmium ion
cellular response to hypoxia
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
heme oxygenase (decyclizing) activity
phospholipase D activity
signal transducer activity
protein binding
enzyme binding
heme binding
protein homodimerization activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0