Search
Search

GALNT9| Gene Symbol: GALNT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122636.1 | NP_001116108.1 | 8 | 1488 | ||
| NM_021808.3 | NP_068580.2 | 4 | 552 | |||
| XM_017019371.1 | XP_016874860.1 | 7 | 1220 | |||
| XM_017019372.1 | XP_016874861.1 | 7 | 1180 | |||
| XM_017019373.1 | XP_016874862.1 | 8 | 1537 | |||
| XM_017019374.1 | XP_016874863.1 | 7 | 1157 | |||
| GenBank mRNA | AB040672.2 | - | 8 | 1372 | ||
| AF458594.1 | - | 4 | 549 | |||
| AJ505960.1 | - | 1 | 94 | |||
| AK054820.1 | - | 5 | 689 | |||
| AK055773.1 | - | 7 | 1145 | |||
| BC093817.1 | - | 3 | 362 | |||
| BC093819.1 | - | 3 | 362 | |||
| BI760629.1 | - | 3 | 624 | |||
| CB154648.1 | - | 2 | 148 | |||
| DR001000.1 | - | 3 | 356 | |||