Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF1A SIRNA Assays ›
Gene Symbol:
KIF1A
Gene Name:
kinesin family member 1A
Gene Aliases:
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location:
Chr.2: 240713764 - 240821025 on Build GRCh38
UniGene ID:
Hs.516802
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224318
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244008.1
NP_001230937.1
37
4091
NM_001320705.1
NP_001307634.1
36
3815
NM_004321.6
NP_004312.2
34
3601
XM_005247022.1
XP_005247079.1
37
4002
XM_005247024.1
XP_005247081.1
36
3975
XM_005247027.1
XP_005247084.1
35
3699
XM_005247028.2
XP_005247085.1
35
3702
XM_006712605.1
XP_006712668.1
36
3975
XM_011511364.1
XP_011509666.1
37
4014
XM_011511365.1
XP_011509667.1
36
3726
XM_011511366.2
XP_011509668.1
27
3074
XM_011511367.1
XP_011509669.1
27
3004
XM_017004383.1
XP_016859872.1
37
4077
XM_017004384.1
XP_016859873.1
37
4089
XM_017004385.1
XP_016859874.1
37
4077
XM_017004386.1
XP_016859875.1
37
4077
XM_017004387.1
XP_016859876.1
36
3801
XM_017004388.1
XP_016859877.1
36
3801
XM_017004389.1
XP_016859878.1
36
3801
XM_017004390.1
XP_016859879.1
35
3699
XM_017004391.1
XP_016859880.1
36
3856
GenBank mRNA
AB209138.1
-
10
900
AB210029.1
-
37
4091
AB290172.1
-
37
4091
BC111780.1
-
34
3569
40037563
BC111799.1
-
34
3569
40037561
KJ534875.1
-
34
3541
U37194.1
-
6
454
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
anterograde axonal transport
motor activity
microtubule motor activity
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline