Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GATAD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300946.1 | NP_001287875.1 | 10 | 1648 | ||
NM_017660.3 | NP_060130.3 | 11 | 1951 | |||
XM_011528105.1 | XP_011526407.1 | 11 | 1837 | |||
XM_017026901.1 | XP_016882390.1 | 10 | 1726 | |||
XM_017026902.1 | XP_016882391.1 | 12 | 1900 | |||
XM_017026903.1 | XP_016882392.1 | 12 | 1990 | |||
XM_017026904.1 | XP_016882393.1 | 12 | 1947 | |||
XM_017026905.1 | XP_016882394.1 | 12 | 1960 | |||
XM_017026906.1 | XP_016882395.1 | 10 | 1762 | |||
XM_017026907.1 | XP_016882396.1 | 13 | 2069 | |||
XM_017026908.1 | XP_016882397.1 | 14 | 2329 | |||
XM_017026909.1 | XP_016882398.1 | 14 | 2280 | |||
XM_017026910.1 | XP_016882399.1 | 12 | 1889 | |||
XM_017026911.1 | XP_016882400.1 | 12 | 2061 | |||
GenBank mRNA | AK000092.1 | - | 3 | 393 | ||
AK125974.1 | - | 13 | 2059 | |||
AK225198.1 | - | 3 | 393 | |||
AK296781.1 | - | 7 | 1209 | |||
AK303482.1 | - | 6 | 1159 | |||
AK315416.1 | - | 10 | 1524 | |||
AL390164.1 | - | 2 | 174 | |||
AY186731.1 | - | 10 | 1639 | |||
BC011684.1 | - | 11 | 1680 | 3953055 | ||
BC012902.1 | - | 4 | 539 |