Search
Search

NAGK| Gene Symbol: NAGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017567.4 | NP_060037.3 | 6 | 951 | ||
| XM_005264412.4 | XP_005264469.1 | 6 | 607 | |||
| XM_005264413.1 | XP_005264470.1 | 6 | 708 | |||
| XM_005264415.2 | XP_005264472.1 | 5 | 819 | |||
| XM_011532957.1 | XP_011531259.1 | 6 | 916 | |||
| XM_017004440.1 | XP_016859929.1 | 6 | 951 | |||
| GenBank mRNA | AJ242910.1 | - | 6 | 562 | ||
| AK001812.1 | - | 6 | 580 | |||
| AK097223.1 | - | 5 | 2369 | |||
| AK222645.1 | - | 6 | 579 | |||
| AK297224.1 | - | 6 | 928 | |||
| AL832837.1 | - | 2 | 3791 | |||
| BC001029.2 | - | 6 | 593 | 3347484 | ||
| BC005371.1 | - | 6 | 583 | 3932248 | ||
| CR457271.1 | - | 6 | 545 | |||