Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUF60 SIRNA Assays ›
Gene Symbol:
PUF60
Gene Name:
poly(U) binding splicing factor 60
Gene Aliases:
FIR, RoBPI, SIAHBP1, VRJS
Chromosome Location:
Chr.8: 143816344 - 143829386 on Build GRCh38
UniGene ID:
Hs.521924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22445
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUF60
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136033.2
NP_001129505.1
8, 9
919
NM_001271096.1
NP_001258025.1
7, 8
846
NM_001271097.1
NP_001258026.1
6, 7
762
NM_001271098.1
NP_001258027.1
8, 9
897
NM_001271099.1
NP_001258028.1
7, 8
813
NM_001271100.1
NP_001258029.1
7, 8
868
NM_014281.4
NP_055096.2
7, 8
849
NM_078480.2
NP_510965.1
8, 9
900
XM_011516929.2
XP_011515231.1
9, 10
1099
XM_011516930.2
XP_011515232.1
8, 9
950
XM_017013234.1
XP_016868723.1
8, 9
2432
XM_017013235.1
XP_016868724.1
8, 9
1002
XM_017013236.1
XP_016868725.1
7, 8
2381
XM_017013237.1
XP_016868726.1
9, 10
1161
XM_017013238.1
XP_016868727.1
9, 10
961
XM_017013239.1
XP_016868728.1
8, 9
927
XM_017013240.1
XP_016868729.1
7, 8
876
GenBank mRNA
AF114818.1
-
8, 9
881
AF190744.1
-
8, 9
809
AF217197.2
-
7, 8
776
AK055941.1
-
7, 8
1503
AK292373.1
-
8, 9
881
AL522753.3
-
8, 9
847
BC008875.2
-
7, 8
815
4138778
BC009734.1
-
7, 8
798
3877563
BC011265.1
-
8, 9
836
4155549
BC011979.1
-
6, 7
715
3863660
BC035435.2
-
7, 8
800
BX418925.2
-
7, 8
798
CR457143.1
-
8, 9
817
CT000277.1
-
7, 8
781
CX163878.1
-
6, 7
720
U51586.1
-
6, 7
763
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
RNA splicing
cell-cell adhesion
nucleotide binding
DNA binding
protein binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0