Search Thermo Fisher Scientific
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020387.3 | NP_065120.2 | 3 | 537 | ||
NR_133653.1 | 2 | 341 | ||||
GenBank mRNA | AF083124.1 | - | 3 | 498 | ||
AF274025.1 | - | 3 | 516 | |||
BC004416.1 | - | 1 | 27 | |||
BC009831.2 | - | 3 | 511 | 3926839 | ||
BC033322.1 | - | 3 | 501 | 4775725 | ||
BP273992.1 | - | 2 | 326 |