Search Thermo Fisher Scientific
Gene Symbol: NLGN4Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164238.1 | NP_001157710.1 | 1 | 556 | ||
NM_014893.4 | NP_055708.3 | 2 | 709 | |||
NR_028319.1 | 2 | 931 | ||||
NR_046355.1 | 2 | 636 | ||||
XM_006724874.2 | XP_006724937.1 | 2 | 881 | |||
XM_011531424.2 | XP_011529726.1 | 2 | 1199 | |||
XM_011531425.2 | XP_011529727.1 | 2 | 971 | |||
XM_011531426.2 | XP_011529728.1 | 2 | 932 | |||
XM_011531427.1 | XP_011529729.1 | 2 | 686 | |||
XM_011531428.1 | XP_011529730.1 | 3 | 867 | |||
XM_011531429.2 | XP_011529731.1 | 2 | 1482 | |||
XM_011531430.2 | XP_011529732.1 | 2 | 820 | |||
XM_017030034.1 | XP_016885523.1 | 2 | 971 | |||
XM_017030035.1 | XP_016885524.1 | 2 | 892 | |||
XM_017030036.1 | XP_016885525.1 | 2 | 686 | |||
XM_017030037.1 | XP_016885526.1 | 2 | 684 | |||
XM_017030038.1 | XP_016885527.1 | 2 | 1482 | |||
XM_017030039.1 | XP_016885528.1 | 1 | 63 | |||
XM_017030041.1 | XP_016885530.1 | 2 | 970 | |||
GenBank mRNA | AF376804.1 | - | 2 | 709 | ||
AK295117.1 | - | 2 | 636 | |||
BC032567.1 | - | 1 | 359 | |||
BC113525.1 | - | 1 | 536 | 8322577 | ||
BC113551.1 | - | 1 | 536 | 8322603 | ||
BX537428.1 | - | 2 | 932 |