Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SF1 SIRNA Assays ›
Gene Symbol:
SF1
Gene Name:
splicing factor 1
Gene Aliases:
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
Chromosome Location:
Chr.11: 64764604 - 64779043 on Build GRCh38
UniGene ID:
Hs.502829
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224782
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178030.1
NP_001171501.1
7, 8
1222
NM_001178031.1
NP_001171502.1
7, 8
1004
NM_004630.3
NP_004621.2
7, 8
1225
NM_201995.2
NP_973724.1
7, 8
1225
NM_201997.2
NP_973726.2
7, 8
1225
NM_201998.2
NP_973727.1
7, 8
1225
NR_033649.1
6, 7
1149
NR_033650.1
6, 7
1149
XM_011545244.1
XP_011543546.1
7, 8
897
XM_011545245.1
XP_011543547.1
7, 8
897
XM_011545246.1
XP_011543548.1
7, 8
897
XM_011545247.1
XP_011543549.1
7, 8
897
XM_011545248.1
XP_011543550.1
7, 8
897
XM_017018244.1
XP_016873733.1
7, 8
897
XM_017018245.1
XP_016873734.1
7, 8
978
XM_017018246.1
XP_016873735.1
7, 8
897
XM_017018247.1
XP_016873736.1
7, 8
940
XM_017018248.1
XP_016873737.1
7, 8
940
XM_017018249.1
XP_016873738.1
7, 8
3390
XM_017018250.1
XP_016873739.1
6, 7
1074
XM_017018251.1
XP_016873740.1
6, 7
929
GenBank mRNA
AK293753.1
-
7, 8
1004
AK296237.1
-
7, 8
896
AK299705.1
-
5, 6
635
AK301803.1
-
6, 7
1074
BC008080.2
-
7, 8
921
2961666
BC008724.2
-
7, 8
921
2961666
BC020217.1
-
7, 8
1124
4869292
BC034451.1
-
7, 8
1150
BC038446.1
-
7, 8
1245
5493676
D26120.1
-
7, 8
1155
L49345.1
-
7, 8
1111
L49380.1
-
7, 8
1111
Y08765.1
-
7, 8
815
Y08766.1
-
7, 8
1067
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
male sex determination
Leydig cell differentiation
negative regulation of smooth muscle cell proliferation
regulation of steroid biosynthetic process
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0