Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACBD4 SIRNA Assays ›
          Gene Symbol:
          ACBD4
          Gene Name:
          acyl-CoA binding domain containing 4
          Gene Aliases:
          HMFT0700
          Chromosome Location:
          Chr.17: 45132600 - 45144181 on Build GRCh38
          UniGene ID:
          Hs.110298
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224830

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACBD4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135705.2 NP_001129177.1 5, 6 759
          NM_001135706.2 NP_001129178.1 5, 6 759
          NM_001135707.2 NP_001129179.1 5, 6 759
          NM_001321352.1 NP_001308281.1 7, 8 883
          NM_001321353.1 NP_001308282.1 7, 8 880
          NM_024722.3 NP_078998.1 4, 5 913
          XM_006722085.2 XP_006722148.1 6, 7 1129
          XM_011525259.2 XP_011523561.1 6, 7 1129
          XM_011525261.2 XP_011523563.1 6, 7 1129
          XM_017025084.1 XP_016880573.1 6, 7 1129
          XM_017025085.1 XP_016880574.1 7, 8 880
          XM_017025086.1 XP_016880575.1 8, 9 1029
          XM_017025087.1 XP_016880576.1 6, 7 1129
          XM_017025088.1 XP_016880577.1 6, 7 1129
          XM_017025089.1 XP_016880578.1 6, 7 1129
          XM_017025090.1 XP_016880579.1 6, 7 1129
          XM_017025091.1 XP_016880580.1 6, 7 1129
          XM_017025092.1 XP_016880581.1 7, 8 1254
          XM_017025093.1 XP_016880582.1 6, 7 1129
          XM_017025094.1 XP_016880583.1 4, 5 873
          XM_017025095.1 XP_016880584.1 6, 7 1129
          XM_017025096.1 XP_016880585.1 6, 7 1129
          XM_017025097.1 XP_016880586.1 6, 7 1129
          XM_017025098.1 XP_016880587.1 7, 8 880
          XM_017025099.1 XP_016880588.1 6, 7 1129
          XM_017025100.1 XP_016880589.1 6, 7 1129
          XM_017025101.1 XP_016880590.1 6, 7 1129
          GenBank mRNA AB073387.1 - 5, 6 671
          AK023384.1 - 4, 5 873
          AK075104.1 - 5, 6 712
          AK296167.1 - 5, 6 746
          AK316359.1 - 5, 6 759
          BC029164.1 - 5, 6 698 5188183
          BC041143.1 - 7, 8 852 5769682
          KU178779.1 - 4, 5 415
          KU178780.1 - 4, 5 415
          KU178781.1 - 4, 5 415
          KU178782.1 - 4, 5 415

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Process(es)

          fatty-acyl-CoA binding
          lipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0