Search
Search

MEGF10| Gene Symbol: MEGF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256545.1 | NP_001243474.1 | 24 | 3503 | ||
| NM_032446.2 | NP_115822.1 | 25 | 3554 | |||
| XM_011543694.1 | XP_011541996.1 | 24 | 3266 | |||
| XM_017009987.1 | XP_016865476.1 | 26 | 3799 | |||
| XM_017009988.1 | XP_016865477.1 | 17 | 2572 | |||
| GenBank mRNA | AB058676.1 | - | 25 | 3395 | ||
| BC152478.1 | - | 25 | 3395 | 8862616 | ||
| CR749437.1 | - | 24 | 3437 | |||