Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320099.1 | NP_001307028.1 | 2, 3 | 624 | ||
NM_003805.4 | NP_003796.1 | 2, 3 | 414 | |||
GenBank mRNA | AB451280.1 | - | 1, 2 | 306 | ||
AB451412.1 | - | 1, 2 | 306 | |||
AK314861.1 | - | 2, 3 | 394 | |||
BC017042.1 | - | 2, 3 | 386 | 3849848 | ||
BC037905.1 | - | 2, 3 | 628 | 5276510 | ||
BT009837.1 | - | 1, 2 | 306 | |||
CR407643.1 | - | 1, 2 | 306 | |||
HY113325.1 | - | 2, 3 | 415 | |||
U79115.1 | - | 2, 3 | 406 | |||
U84388.1 | - | 2, 3 | 343 |