Search
Search

CCNH| Gene Symbol: CCNH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199189.1 | NP_001186118.1 | 6, 7 | 1122 | ||
| NM_001239.3 | NP_001230.1 | 6, 7 | 1008 | |||
| XM_005248627.3 | XP_005248684.1 | 6, 7 | 846 | |||
| XM_005248629.4 | XP_005248686.1 | 6, 7 | 1184 | |||
| XM_011543706.2 | XP_011542008.1 | 6, 7 | 846 | |||
| XM_017010014.1 | XP_016865503.1 | 6, 7 | 1148 | |||
| GenBank mRNA | AB209342.1 | - | 5, 6 | 600 | ||
| AK094534.1 | - | 2, 3 | 285 | |||
| AK291720.1 | - | 6, 7 | 861 | |||
| BC005280.1 | - | 6, 7 | 864 | 3930062 | ||
| BC016705.1 | - | 6, 7 | 859 | 4082151 | ||
| BC016823.1 | - | 6, 7 | 855 | 4102297 | ||
| BC022351.1 | - | 6, 7 | 843 | 4277337 | ||
| BX537673.1 | - | 6, 7 | 1123 | |||
| CR407658.1 | - | 6, 7 | 769 | |||
| U11791.1 | - | 6, 7 | 829 | |||
| U12685.1 | - | 6, 7 | 781 | |||