Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRXN1 SIRNA Assays ›
          Gene Symbol:
          NRXN1
          Gene Name:
          neurexin 1
          Gene Aliases:
          Hs.22998, PTHSL2, SCZD17
          Chromosome Location:
          Chr.2: 49918503 - 51032536 on Build GRCh38
          UniGene ID:
          Hs.637685
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s225122

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 388.00
          Your Price
          Online offer: 374.65
          374.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRXN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135659.1 NP_001129131.1 22 5597
          NM_004801.4 NP_004792.1 20 5387
          NM_138735.2 NP_620072.1 4 1612
          XM_005264642.3 XP_005264699.1 21 5498
          XM_005264643.3 XP_005264700.1 20 5453
          XM_006712137.3 XP_006712200.1 20 5408
          XM_006712140.3 XP_006712203.2 22 5525
          XM_011533167.2 XP_011531469.1 21 5498
          XM_011533171.2 XP_011531473.1 21 5477
          XM_011533172.2 XP_011531474.1 20 5471
          XM_011533174.2 XP_011531476.1 20 5453
          XM_011533175.2 XP_011531477.1 19 5441
          XM_011533177.2 XP_011531479.1 20 5408
          XM_011533178.2 XP_011531480.1 18 5408
          XM_011533180.2 XP_011531482.1 21 5498
          XM_011533183.1 XP_011531485.1 15 3180
          XM_017005303.1 XP_016860792.1 22 5525
          XM_017005304.1 XP_016860793.1 22 5522
          XM_017005305.1 XP_016860794.1 22 5525
          XM_017005306.1 XP_016860795.1 21 5513
          XM_017005307.1 XP_016860796.1 21 5507
          XM_017005308.1 XP_016860797.1 22 5504
          XM_017005309.1 XP_016860798.1 21 5498
          XM_017005310.1 XP_016860799.1 20 5495
          XM_017005311.1 XP_016860800.1 21 5480
          XM_017005312.1 XP_016860801.1 21 5474
          XM_017005313.1 XP_016860802.1 21 5477
          XM_017005314.1 XP_016860803.1 19 5465
          XM_017005315.1 XP_016860804.1 20 5471
          XM_017005316.1 XP_016860805.1 20 5462
          XM_017005317.1 XP_016860806.1 20 5465
          XM_017005318.1 XP_016860807.1 20 5453
          XM_017005319.1 XP_016860808.1 20 5450
          XM_017005320.1 XP_016860809.1 20 5450
          XM_017005321.1 XP_016860810.1 21 5435
          XM_017005322.1 XP_016860811.1 21 5435
          XM_017005323.1 XP_016860812.1 17 5411
          XM_017005324.1 XP_016860813.1 19 5381
          XM_017005325.1 XP_016860814.1 19 5381
          XM_017005326.1 XP_016860815.1 19 5369
          XM_017005327.1 XP_016860816.1 19 5363
          XM_017005328.1 XP_016860817.1 18 5336
          XM_017005329.1 XP_016860818.1 22 5525
          XM_017005330.1 XP_016860819.1 16 5321
          XM_017005331.1 XP_016860820.1 16 5306
          XM_017005332.1 XP_016860821.1 15 5276
          XM_017005333.1 XP_016860822.1 15 5276
          XM_017005334.1 XP_016860823.1 18 3346
          XM_017005335.1 XP_016860824.1 5 6692
          XM_017005336.1 XP_016860825.1 5 6693
          XM_017005337.1 XP_016860826.1 4 6602
          GenBank mRNA AB011150.2 - 19 4159
          AB035356.1 - 19 4102
          BC125179.1 - 5 668
          BC125180.1 - 5 1031
          BC150247.1 - 19 4180 9007472
          EF539882.1 - 22 5460

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_021767, M96375, M96374.
          This siRNA also targets the following transcripts in Mouse: NM_177284, NM_020252, AK017578, BC079893, AB093249.

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron cell-cell adhesion
          chemical synaptic transmission
          neurotransmitter secretion
          axon guidance
          synapse assembly
          learning
          adult behavior
          social behavior
          vocal learning
          neuromuscular process controlling balance
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          prepulse inhibition
          regulation of insulin secretion involved in cellular response to glucose stimulus
          vocalization behavior
          positive regulation of synapse maturation
          postsynaptic membrane assembly
          gephyrin clustering involved in postsynaptic density assembly
          neuroligin clustering involved in postsynaptic membrane assembly
          postsynaptic density protein 95 clustering
          positive regulation of excitatory postsynaptic potential
          regulation of grooming behavior
          receptor activity
          calcium channel regulator activity
          calcium ion binding
          protein binding
          acetylcholine receptor binding
          cell adhesion molecule binding
          neuroligin family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline