Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318842.1 | NP_001305771.1 | 2 | 633 | ||
NM_001318847.1 | NP_001305776.1 | 2 | 633 | |||
NM_001318848.1 | NP_001305777.1 | 2 | 633 | |||
NM_006586.4 | NP_006577.2 | 2 | 633 | |||
NR_134880.1 | 2 | 633 | ||||
NR_134881.1 | 3 | 1826 | ||||
NR_134885.1 | 2 | 730 | ||||
GenBank mRNA | AK090425.1 | - | 1 | 82 | ||
AK124196.1 | - | 3 | 1825 | |||
AY358960.1 | - | 2 | 528 | |||
BC004423.1 | - | 2 | 579 | 2819660 | ||
BC008898.2 | - | 2 | 260 | 2959532 | ||
BC008961.2 | - | 2 | 260 | 2959532 | ||
BG675987.1 | - | 2 | 227 | |||
BQ063472.1 | - | 2 | 239 | |||
BQ680146.1 | - | 2 | 252 | |||
CK429647.1 | - | 2 | 246 | |||
CN302713.1 | - | 2 | 371 | |||
U80744.1 | - | 2 | 579 |