Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FBXL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152441.2 | NP_689654.1 | 1 | 558 | ||
XM_017018875.1 | XP_016874364.1 | 1 | 582 | |||
XM_017018876.1 | XP_016874365.1 | 1 | 582 | |||
GenBank mRNA | BC028132.1 | - | 1 | 558 | 5205284 |