Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP54 SIRNA Assays ›
Gene Symbol:
USP54
Gene Name:
ubiquitin specific peptidase 54
Gene Aliases:
C10orf29, bA137L10.3, bA137L10.4
Chromosome Location:
Chr.10: 73497538 - 73626844 on Build GRCh38
UniGene ID:
Hs.657355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225771
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP54
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320437.1
NP_001307366.1
13, 14
1988
NM_152586.3
NP_689799.3
13, 14
2086
NR_135249.1
16, 17
2789
NR_135250.1
11, 12
1784
XM_005269582.2
XP_005269639.1
12, 13
5164
XM_011539368.2
XP_011537670.1
13, 14
5230
XM_017015759.1
XP_016871248.1
15, 16
4659
XM_017015760.1
XP_016871249.1
15, 16
3561
XM_017015761.1
XP_016871250.1
15, 16
4399
XM_017015762.1
XP_016871251.1
15, 16
3569
XM_017015763.1
XP_016871252.1
15, 16
2863
XM_017015764.1
XP_016871253.1
15, 16
2926
XM_017015765.1
XP_016871254.1
16, 17
3179
XM_017015766.1
XP_016871255.1
15, 16
4785
XM_017015767.1
XP_016871256.1
15, 16
4444
XM_017015768.1
XP_016871257.1
15, 16
2615
XM_017015769.1
XP_016871258.1
15, 16
2223
XM_017015770.1
XP_016871259.1
15, 16
2884
XM_017015771.1
XP_016871260.1
14, 15
5401
XM_017015772.1
XP_016871261.1
14, 15
5407
XM_017015773.1
XP_016871262.1
13, 14
5341
XM_017015774.1
XP_016871263.1
14, 15
4327
XM_017015775.1
XP_016871264.1
15, 16
4205
XM_017015776.1
XP_016871265.1
14, 15
4710
XM_017015777.1
XP_016871266.1
14, 15
4369
XM_017015778.1
XP_016871267.1
14, 15
5407
XM_017015779.1
XP_016871268.1
14, 15
5407
XM_017015780.1
XP_016871269.1
14, 15
5407
XM_017015781.1
XP_016871270.1
13, 14
5236
XM_017015782.1
XP_016871271.1
15, 16
3107
XM_017015783.1
XP_016871272.1
14, 15
4034
XM_017015784.1
XP_016871273.1
13, 14
1980
XM_017015785.1
XP_016871274.1
14, 15
5407
XM_017015786.1
XP_016871275.1
14, 15
2597
GenBank mRNA
AK126497.1
-
3, 4
757
AK127213.1
-
1, 2
217
AK127267.1
-
11, 12
1784
BX649087.1
-
14, 15
2629
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
protein deubiquitination
protein binding
thiol-dependent ubiquitinyl hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0