Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD6 SIRNA Assays ›
          Gene Symbol:
          ANKRD6
          Gene Name:
          ankyrin repeat domain 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 89433178 - 89633834 on Build GRCh38
          UniGene ID:
          Hs.702213
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22578

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242809.1 NP_001229738.1 2 417
          NM_001242811.1 NP_001229740.1 2 800
          NM_001242813.1 NP_001229742.1 2 417
          NM_001242814.1 NP_001229743.1 1 215
          NM_014942.4 NP_055757.3 2 417
          XM_005248680.3 XP_005248737.1 2 2250
          XM_005248681.2 XP_005248738.1 2 518
          XM_011535598.2 XP_011533900.1 2 386
          XM_011535599.2 XP_011533901.1 2 386
          XM_011535600.2 XP_011533902.1 2 386
          XM_017010495.1 XP_016865984.1 2 386
          XM_017010496.1 XP_016865985.1 2 550
          XM_017010497.1 XP_016865986.1 1 219
          XM_017010498.1 XP_016865987.1 2 386
          XM_017010501.1 XP_016865990.1 2 386
          XM_017010502.1 XP_016865991.1 2 2067
          XM_017010503.1 XP_016865992.1 2 2071
          XM_017010504.1 XP_016865993.1 1 219
          XM_017010505.1 XP_016865994.1 2 386
          XM_017010506.1 XP_016865995.1 1 219
          XM_017010507.1 XP_016865996.1 1 219
          XM_017010508.1 XP_016865997.1 2 2062
          XM_017010509.1 XP_016865998.1 1 219
          XM_017010510.1 XP_016865999.1 2 373
          XM_017010511.1 XP_016866000.1 2 2076
          XM_017010512.1 XP_016866001.1 2 386
          XM_017010513.1 XP_016866002.1 2 518
          XM_017010514.1 XP_016866003.1 2 386
          XM_017010515.1 XP_016866004.1 1 219
          GenBank mRNA AB023174.1 - 2 244
          AK096867.1 - 1 77
          AK295731.1 - 2 717
          AK307792.1 - 2 386
          BC042173.1 - 2 382 5454509
          DN999929.1 - 2 426

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001134969, XM_006237976, XM_008763640, XM_006237978, XM_006237977, and others.
          This siRNA also targets the following transcripts in Mouse: NM_080471, NM_001012450, NM_001012451, AK173070, AK083265, and others.

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase inhibitor kinase modulator non-receptor serine/threonine protein kinase ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s)

          positive regulation of JNK cascade
          negative regulation of canonical Wnt signaling pathway
          positive regulation of Wnt signaling pathway, planar cell polarity pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0