Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR8B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005195.1 | NP_001005195.1 | 1 | 319 | ||
GenBank mRNA | BC136863.1 | - | 1 | 344 | 9020854 | |
BC136864.1 | - | 1 | 344 | 9020855 |