Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318818.1 | NP_001305747.1 | 7 | 632 | ||
NM_001318820.1 | NP_001305749.1 | 6 | 690 | |||
NM_001318821.1 | NP_001305750.1 | 7 | 798 | |||
NM_001318822.1 | NP_001305751.1 | 5 | 613 | |||
NM_001318823.1 | NP_001305752.1 | 6 | 721 | |||
NM_001318824.1 | NP_001305753.1 | 7 | 601 | |||
NM_182612.3 | NP_872418.1 | 8 | 709 | |||
NR_134867.1 | 7 | 740 | ||||
NR_134868.1 | 8 | 803 | ||||
XM_017017664.1 | XP_016873153.1 | 7 | 824 | |||
XM_017017667.1 | XP_016873156.1 | 6 | 730 | |||
XM_017017668.1 | XP_016873157.1 | 6 | 812 | |||
GenBank mRNA | AK091602.1 | - | 8 | 691 | ||
AK092816.1 | - | 7 | 586 | |||
AK128653.1 | - | 5 | 958 | |||
AK293559.1 | - | 7 | 617 | |||
AK309153.1 | - | 5 | 932 | |||
BC112145.1 | - | 8 | 690 | 8327613 | ||
BC143415.1 | - | 5 | 594 | |||
BC143416.1 | - | 6 | 671 | 9051924 | ||
BC143418.1 | - | 7 | 779 | 9051926 | ||
BC143419.1 | - | 7 | 613 | |||
BC143420.1 | - | 7 | 582 | 9051928 | ||
BC143421.1 | - | 8 | 784 | |||
BC143422.1 | - | 6 | 702 | |||
BC143423.1 | - | 7 | 721 |