Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 10 | 1060 | ||
XM_011544605.2 | XP_011542907.1 | 10 | 945 | |||
XM_011544606.2 | XP_011542908.1 | 10 | 996 | |||
XM_011544607.2 | XP_011542909.1 | 10 | 958 | |||
XM_017013718.1 | XP_016869207.1 | 9 | 811 | |||
XM_017013719.1 | XP_016869208.1 | 9 | 781 | |||
XM_017013720.1 | XP_016869209.1 | 9 | 896 | |||
GenBank mRNA | AF286164.1 | - | 10 | 985 | ||
AK291842.1 | - | 10 | 1004 | |||
AK309020.1 | - | 3 | 177 | |||
DC332022.1 | - | 3 | 177 |