Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST5 SIRNA Assays ›
          Gene Symbol:
          ST5
          Gene Name:
          suppression of tumorigenicity 5
          Gene Aliases:
          DENND2B, HTS1, p126
          Chromosome Location:
          Chr.11: 8693352 - 8910951 on Build GRCh38
          UniGene ID:
          Hs.117715
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s227457

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005418.3 NP_005409.3 10, 11 2324
          NM_139157.2 NP_631896.1 6, 7 792
          NM_213618.1 NP_998783.1 7, 8 2124
          XM_005253077.1 XP_005253134.1 7, 8 2039
          XM_005253083.2 XP_005253140.1 6, 7 934
          XM_011520309.1 XP_011518611.1 7, 8 2039
          XM_011520310.1 XP_011518612.1 7, 8 2451
          XM_011520311.2 XP_011518613.1 7, 8 2433
          XM_011520312.1 XP_011518614.1 11, 12 2398
          XM_011520313.1 XP_011518615.1 10, 11 2284
          XM_011520314.1 XP_011518616.1 9, 10 2176
          XM_011520315.1 XP_011518617.1 9, 10 2272
          XM_011520316.1 XP_011518618.1 8, 9 2202
          XM_011520317.1 XP_011518619.1 7, 8 2143
          XM_011520318.1 XP_011518620.1 11, 12 2398
          XM_011520319.2 XP_011518621.1 7, 8 1991
          XM_011520320.1 XP_011518622.1 7, 8 2039
          XM_011520321.2 XP_011518623.1 7, 8 2039
          XM_011520322.1 XP_011518624.1 7, 8 2039
          XM_011520323.2 XP_011518625.2 6, 7 862
          XM_011520324.2 XP_011518626.2 6, 7 862
          XM_011520325.1 XP_011518627.1 6, 7 934
          XM_011520326.1 XP_011518628.1 10, 11 1138
          XM_011520327.1 XP_011518629.1 6, 7 827
          XM_011520328.1 XP_011518630.1 6, 7 859
          XM_011520329.1 XP_011518631.1 10, 11 1138
          XM_017018182.1 XP_016873671.1 8, 9 2157
          XM_017018183.1 XP_016873672.1 8, 9 2134
          XM_017018184.1 XP_016873673.1 10, 11 2293
          XM_017018185.1 XP_016873674.1 6, 7 2089
          XM_017018186.1 XP_016873675.1 6, 7 2089
          XM_017018187.1 XP_016873676.1 6, 7 798
          GenBank mRNA AK127763.1 - 4, 5 1179
          AK293245.1 - 4, 5 557
          AK302824.1 - 6, 7 771
          AK312305.1 - 7, 8 2090
          AK312758.1 - 6, 7 1134
          BC036655.1 - 10, 11 2329 4837231
          BX647431.1 - 7, 8 2124
          S45936.1 - 3, 4 462
          U15131.1 - 7, 8 2052
          U15779.1 - 3, 4 421
          U15780.1 - 6, 7 792

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          Rab guanyl-nucleotide exchange factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dhnrf:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0