Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018452.5 | NP_060922.2 | 2 | 889 | ||
GenBank mRNA | AF217510.1 | - | 2 | 231 | ||
AK000891.1 | - | 2 | 137 | |||
BC029130.1 | - | 2 | 157 | 5182086 | ||
BC146893.1 | - | 2 | 199 | 9056821 | ||
BC146895.1 | - | 2 | 199 | 9056823 | ||
DA684766.1 | - | 2 | 392 |