Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012308.2 | NP_036440.1 | 5 | 1151 | ||
NR_027473.1 | 5 | 1151 | ||||
XM_011544860.2 | XP_011543162.1 | 5 | 1210 | |||
XM_017017391.1 | XP_016872880.1 | 5 | 457 | |||
XM_017017392.1 | XP_016872881.1 | 4 | 268 | |||
GenBank mRNA | AB023221.2 | - | 5 | 468 | ||
AK024505.1 | - | 5 | 393 | |||
BC047486.1 | - | 5 | 1153 | 5296445 | ||
BC064360.1 | - | 5 | 566 | 5534384 | ||
JQ710741.1 | - | 4 | 335 | |||
JQ710742.1 | - | 5 | 457 | |||
JQ710743.1 | - | 4 | 335 | |||
JQ710744.1 | - | 6 | 529 |