Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: PALLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166108.1 | NP_001159580.1 | 17 | 2954 | ||
NM_001166109.1 | NP_001159581.1 | 15 | 2068 | |||
NM_001166110.1 | NP_001159582.1 | 8 | 1567 | |||
NM_016081.3 | NP_057165.3 | 16 | 2903 | |||
XM_005262861.4 | XP_005262918.1 | 18 | 3653 | |||
XM_005262866.2 | XP_005262923.1 | 17 | 3203 | |||
XM_011531768.2 | XP_011530070.1 | 19 | 3907 | |||
XM_011531769.2 | XP_011530071.1 | 18 | 3856 | |||
XM_011531770.2 | XP_011530072.1 | 19 | 3907 | |||
XM_011531771.2 | XP_011530073.1 | 17 | 3634 | |||
XM_011531772.2 | XP_011530074.1 | 16 | 3535 | |||
XM_011531773.1 | XP_011530075.1 | 18 | 3235 | |||
XM_011531774.1 | XP_011530076.1 | 17 | 3184 | |||
XM_011531775.1 | XP_011530077.1 | 17 | 2588 | |||
XM_017007910.1 | XP_016863399.1 | 18 | 3856 | |||
GenBank mRNA | AB023209.1 | - | 8 | 1531 | ||
AF077041.1 | - | 4 | 1017 | |||
AF151909.1 | - | 4 | 923 | |||
AF464873.1 | - | 16 | 2903 | |||
AK000845.1 | - | 4 | 492 | |||
AK025843.1 | - | 4 | 494 | |||
AK095512.1 | - | 15 | 2030 | |||
AY211921.1 | - | 7 | 753 | |||
BC013867.2 | - | 7 | 747 | |||
BC140850.1 | - | 16 | 2865 | 9021718 | ||
BC144666.1 | - | 17 | 2916 | 9053197 |