Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1D SIRNA Assays ›
          Gene Symbol:
          CACNA1D
          Gene Name:
          calcium voltage-gated channel subunit alpha1 D
          Gene Aliases:
          CACH3, CACN4, CACNL1A2, CCHL1A2, Cav1.3, PASNA, SANDD
          Chromosome Location:
          Chr.3: 53495049 - 53813151 on Build GRCh38
          UniGene ID:
          Hs.476358
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2287

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000720.3 NP_000711.1 33 4150
          NM_001128839.2 NP_001122311.1 31 4045
          NM_001128840.2 NP_001122312.1 32 4090
          XM_005265448.3 XP_005265505.1 31 3927
          XM_011534094.2 XP_011532396.1 34 4242
          XM_011534096.2 XP_011532398.1 31 4053
          XM_011534097.2 XP_011532399.1 32 4709
          XM_011534099.2 XP_011532401.1 30 3692
          XM_011534100.2 XP_011532402.1 32 4137
          XM_017007137.1 XP_016862626.1 34 4242
          XM_017007138.1 XP_016862627.1 34 4239
          XM_017007139.1 XP_016862628.1 34 4242
          XM_017007140.1 XP_016862629.1 33 4182
          XM_017007141.1 XP_016862630.1 33 4182
          XM_017007142.1 XP_016862631.1 33 4158
          XM_017007143.1 XP_016862632.1 33 4158
          XM_017007144.1 XP_016862633.1 33 4158
          XM_017007145.1 XP_016862634.1 32 4113
          GenBank mRNA AB209171.1 - 27 3051
          AK294585.1 - 6 680
          EU363339.1 - 31 3927
          M76558.1 - 32 4482
          M83566.1 - 33 4150

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          adenylate cyclase-modulating G-protein coupled receptor signaling pathway
          sensory perception of sound
          regulation of insulin secretion
          positive regulation of calcium ion transport
          regulation of atrial cardiac muscle cell membrane repolarization
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during cardiac muscle cell action potential
          membrane depolarization during SA node cell action potential
          regulation of heart rate by cardiac conduction
          regulation of potassium ion transmembrane transporter activity
          regulation of potassium ion transmembrane transport
          voltage-gated calcium channel activity
          high voltage-gated calcium channel activity
          ankyrin binding
          metal ion binding
          alpha-actinin binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved SA node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0