Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEGFA SIRNA Assays ›
Gene Symbol:
VEGFA
Gene Name:
vascular endothelial growth factor A
Gene Aliases:
MVCD1, VEGF, VPF
Chromosome Location:
Chr.6: 43770209 - 43786487 on Build GRCh38
UniGene ID:
Hs.73793
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228861
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEGFA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025366.2
NP_001020537.2
2
1145
NM_001025367.2
NP_001020538.2
2
1145
NM_001025368.2
NP_001020539.2
2
1145
NM_001025369.2
NP_001020540.2
2
1145
NM_001025370.2
NP_001020541.2
2
1145
NM_001033756.2
NP_001028928.1
2
1145
NM_001171622.1
NP_001165093.1
2
1145
NM_001171623.1
NP_001165094.1
2
1145
NM_001171624.1
NP_001165095.1
2
1145
NM_001171625.1
NP_001165096.1
2
1145
NM_001171626.1
NP_001165097.1
2
1145
NM_001171627.1
NP_001165098.1
2
1145
NM_001171628.1
NP_001165099.1
2
1145
NM_001171629.1
NP_001165100.1
2
1145
NM_001171630.1
NP_001165101.1
2
1145
NM_001204384.1
NP_001191313.1
2
1145
NM_001204385.1
NP_001191314.1
2
1145
NM_001287044.1
NP_001273973.1
2
158
NM_001317010.1
NP_001303939.1
2
1145
NM_003376.5
NP_003367.4
2
1145
GenBank mRNA
AB021221.1
-
2
107
AB209485.1
-
1
1210
AB451322.1
-
2
107
AB451451.1
-
2
107
AF022375.1
-
2
808
AF091352.1
-
2
107
AF214570.1
-
2
107
AF430806.1
-
2
107
AF486837.1
-
2
107
AJ010438.1
-
2
107
AY263145.1
-
2
107
AY500353.1
-
2
107
AY766116.1
-
2
107
BC065522.1
-
2
572
BU153227.1
-
2
582
BX442364.2
-
2
151
DQ229900.1
-
2
107
HY066915.1
-
2
158
KT581010.1
-
1
29
KT581011.1
-
2
107
M27281.1
-
2
263
M32977.1
-
2
163
S85192.1
-
2
175
X62568.1
-
2
123
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
ovarian follicle development
patterning of blood vessels
vasculogenesis
response to hypoxia
in utero embryonic development
kidney development
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
positive regulation of neuroblast proliferation
positive regulation of mesenchymal cell proliferation
positive regulation of receptor internalization
platelet degranulation
positive regulation of leukocyte migration
heart morphogenesis
outflow tract morphogenesis
coronary vein morphogenesis
nervous system development
mesoderm development
lactation
positive regulation of cell proliferation
regulation of cell shape
positive regulation of endothelial cell migration
positive regulation of gene expression
monocyte differentiation
macrophage differentiation
lung development
positive regulation of cell migration
regulation of cGMP metabolic process
epithelial cell differentiation
positive regulation of vascular endothelial growth factor receptor signaling pathway
post-embryonic camera-type eye development
positive regulation of protein complex assembly
positive regulation of protein autophosphorylation
activation of protein kinase activity
positive regulation of CREB transcription factor activity
positive regulation of peptidyl-serine phosphorylation
tube formation
endothelial cell chemotaxis
cellular response to vascular endothelial growth factor stimulus
lymph vessel morphogenesis
positive regulation of endothelial cell chemotaxis by VEGF-activated vascular endothelial growth factor receptor signaling pathway
positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway
VEGF-activated neuropilin signaling pathway
growth
eye photoreceptor cell development
positive regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of apoptotic process
surfactant homeostasis
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAP kinase activity
positive regulation of blood vessel endothelial cell migration
positive regulation of angiogenesis
positive regulation of cell adhesion
positive regulation of transcription from RNA polymerase II promoter
vascular endothelial growth factor receptor signaling pathway
cell maturation
camera-type eye morphogenesis
cardiac muscle fiber development
branching morphogenesis of an epithelial tube
positive regulation of axon extension involved in axon guidance
artery morphogenesis
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive chemotaxis
positive regulation of positive chemotaxis
induction of positive chemotaxis
positive regulation of cellular component movement
positive regulation of cell division
positive regulation of focal adhesion assembly
primitive erythrocyte differentiation
mammary gland alveolus development
cardiac vascular smooth muscle cell development
coronary artery morphogenesis
regulation of transcription from RNA polymerase II promoter in response to hypoxia
positive regulation of transcription from RNA polymerase II promoter in response to hypoxia
cellular response to hypoxia
dopaminergic neuron differentiation
commissural neuron axon guidance
positive regulation of protein kinase C signaling
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of branching involved in ureteric bud morphogenesis
regulation of retinal ganglion cell axon guidance
positive regulation of peptidyl-tyrosine autophosphorylation
positive regulation of p38MAPK cascade
positive regulation of histone deacetylase activity
positive regulation of retinal ganglion cell axon guidance
positive regulation of protein kinase D signaling
fibronectin binding
cytokine activity
platelet-derived growth factor receptor binding
vascular endothelial growth factor receptor binding
protein binding
growth factor activity
heparin binding
chemoattractant activity
identical protein binding
protein homodimerization activity
vascular endothelial growth factor receptor 1 binding
vascular endothelial growth factor receptor 2 binding
protein heterodimerization activity
receptor agonist activity
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-x54js:80/100.66.79.231:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0