Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FNBP1 SIRNA Assays ›
          Gene Symbol:
          FNBP1
          Gene Name:
          formin binding protein 1
          Gene Aliases:
          FBP17
          Chromosome Location:
          Chr.9: 129887187 - 130053846 on Build GRCh38
          UniGene ID:
          Hs.189409
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22914

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FNBP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015033.2 NP_055848.1 8 994
          XM_005251815.2 XP_005251872.1 8 967
          XM_005251820.2 XP_005251877.1 8 967
          XM_005251821.2 XP_005251878.1 8 967
          XM_005251822.2 XP_005251879.1 8 967
          XM_005251823.2 XP_005251880.1 8 967
          XM_005251824.2 XP_005251881.1 8 967
          XM_005251825.2 XP_005251882.1 8 967
          XM_005251826.2 XP_005251883.1 8 967
          XM_005251827.2 XP_005251884.1 8 967
          XM_005251828.2 XP_005251885.1 8 967
          XM_005251829.2 XP_005251886.1 8 967
          XM_005251830.2 XP_005251887.1 8 967
          XM_005251831.2 XP_005251888.1 8 967
          XM_005251832.2 XP_005251889.1 8 967
          XM_005251833.2 XP_005251890.1 8 967
          XM_005251834.2 XP_005251891.1 8 967
          XM_006717016.2 XP_006717079.1 8 967
          XM_011518399.2 XP_011516701.1 8 967
          XM_011518400.2 XP_011516702.1 8 967
          XM_011518401.1 XP_011516703.1 8 967
          XM_011518402.1 XP_011516704.1 8 967
          XM_017014487.1 XP_016869976.1 8 967
          XM_017014488.1 XP_016869977.1 8 967
          XM_017014489.1 XP_016869978.1 8 967
          XM_017014490.1 XP_016869979.1 8 967
          XM_017014491.1 XP_016869980.1 8 967
          XM_017014492.1 XP_016869981.1 8 1253
          XM_017014493.1 XP_016869982.1 8 967
          XM_017014494.1 XP_016869983.1 8 967
          GenBank mRNA AB011126.1 - 8 948
          AF265550.1 - 8 963
          AK000975.1 - 7 671
          AK001616.1 - 7 662
          AK023681.1 - 8 967
          BC062463.1 - 8 909
          BC101755.1 - 8 836 8069261
          BC143513.1 - 8 836 9052022
          BC143514.1 - 8 836 9052023
          BC143515.1 - 8 836 9052024

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          vesicle-mediated transport
          positive regulation of GTPase activity
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          lipid binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hdb68:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0