Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPIAL4G SIRNA Assays ›
          Gene Symbol:
          PPIAL4G LOC105371242 PPIA PPIAL4A PPIAL4C PPIAL4D PPIAL4E PPIAL4F
          Gene Name:
          peptidylprolyl isomerase A like 4G
          peptidyl-prolyl cis-trans isomerase A-like 4C
          peptidylprolyl isomerase A
          peptidylprolyl isomerase A like 4A
          peptidylprolyl isomerase A like 4C
          peptidylprolyl isomerase A like 4D
          peptidylprolyl isomerase A like 4E
          peptidylprolyl isomerase A like 4F
          Gene Aliases:
          COAS-2
          -
          CYPA, CYPH, HEL-S-69p
          COAS-2, COAS2, PPIAL4, PPIAL4B
          COAS-2
          -
          COAS2
          -
          Chromosome Location:
          Chr.1: 148482548 - 148483285 on Build GRCh38
          Chromosome Location:
          Chr.1: 146342335 - 146344907 on Build GRCh38
          Chromosome Location:
          Chr.7: 44796636 - 44803123 on Build GRCh38
          Chromosome Location:
          Chr.1: 120889746 - 120890530 on Build GRCh38
          Chromosome Location:
          Chr.1: 149583823 - 149584607 on Build GRCh38
          Chromosome Location:
          Chr.1: 145241415 - 145242199 on Build GRCh38
          Chromosome Location:
          Chr.1: 144372875 - 144373659 on Build GRCh38
          Chromosome Location:
          Chr.1: 144592868 - 144593652 on Build GRCh38
          UniGene ID:
          Hs.534674
          -
          Hs.356331
          Hs.573713
          Hs.534674
          Hs.730589
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s229589

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPIAL4G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123068.1 NP_001116540.1 1 690
          Gene Symbol: LOC105371242
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011510267.2 XP_011508569.1 1 849
          Gene Symbol: PPIA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300981.1 NP_001287910.1 6 902
          NM_021130.4 NP_066953.1 5 745
          GenBank mRNA AK026569.1 - 1 700
          AK130101.1 - 5 701
          AK290085.1 - 5 701
          AK290851.1 - 5 703
          AK293003.1 - 6 858
          AK297340.1 - 6 592
          BC000689.2 - 1 678 3349335
          BC003026.1 - 1 684 2823490
          BC005320.1 - 1 700 3935025
          BC007104.1 - 1 701 4281376
          BC013915.1 - 1 705 3684721
          BC018843.2 - 1 735
          BC073992.1 - 1 690 6161389
          BC093076.1 - 6 768 30414589
          BC106030.1 - 1 692 2986147
          BM806224.1 - 6 756
          BM909285.1 - 422
          CD014106.1 - 6 858
          EU794650.1 - 1 692
          Y00052.1 - 5 670
          Gene Symbol: PPIAL4A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143883.2 NP_001137355.1 1 732
          Gene Symbol: PPIAL4C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135789.3 NP_001129261.2 1 732
          GenBank mRNA AK123006.1 - 2 1083
          Gene Symbol: PPIAL4D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164261.1 NP_001157733.1 1 732
          Gene Symbol: PPIAL4E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144032.2 NP_001137504.2 1 732
          Gene Symbol: PPIAL4F
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164262.2 NP_001157734.2 1 732

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          protein folding
          RNA-dependent DNA biosynthetic process
          viral life cycle
          uncoating of virus
          fusion of virus membrane with host plasma membrane
          virion assembly
          viral release from host cell
          entry into host cell
          lipid particle organization
          regulation of viral genome replication
          positive regulation of viral genome replication
          positive regulation of protein secretion
          leukocyte migration
          establishment of integrated proviral latency
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          peptide binding
          poly(A) RNA binding
          virion binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0