Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 5 | 669 | ||
NM_001111113.1 | NP_001104583.1 | 3 | 516 | |||
NM_001312647.1 | NP_001299576.1 | 5 | 581 | |||
NM_001312648.1 | NP_001299577.1 | 4 | 575 | |||
NM_005339.4 | NP_005330.1 | 5 | 669 | |||
GenBank mRNA | AB022435.1 | - | 5 | 471 | ||
AB022436.1 | - | 3 | 318 | |||
AK093533.1 | - | 5 | 490 | |||
AK291454.1 | - | 3 | 421 | |||
AK307130.1 | - | 4 | 480 | |||
AK315524.1 | - | 5 | 595 | |||
BC022804.1 | - | 5 | 545 | 4893778 | ||
BC050600.1 | - | 5 | 523 | 6502901 | ||
CX786829.1 | - | 5 | 508 | |||
U58522.1 | - | 5 | 388 |