Search
Search

RUNDC3B| Gene Symbol: RUNDC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134405.1 | NP_001127877.1 | 6 | 1017 | ||
| NM_001134406.1 | NP_001127878.1 | 6 | 1017 | |||
| NM_138290.2 | NP_612147.1 | 7 | 1068 | |||
| XM_005250156.2 | XP_005250213.1 | 6 | 1017 | |||
| XM_005250158.2 | XP_005250215.1 | 4 | 492 | |||
| XM_011515826.1 | XP_011514128.1 | 7 | 1068 | |||
| XM_011515827.1 | XP_011514129.1 | 7 | 1068 | |||
| XM_011515828.2 | XP_011514130.1 | 7 | 551 | |||
| XM_011515829.2 | XP_011514131.1 | 6 | 685 | |||
| XM_017011761.1 | XP_016867250.1 | 7 | 755 | |||
| XM_017011762.1 | XP_016867251.1 | 8 | 680 | |||
| XM_017011763.1 | XP_016867252.1 | 5 | 494 | |||
| XM_017011764.1 | XP_016867253.1 | 4 | 492 | |||
| XM_017011765.1 | XP_016867254.1 | 7 | 667 | |||
| XM_017011766.1 | XP_016867255.1 | 7 | 946 | |||
| GenBank mRNA | AK055233.1 | - | 7 | 1066 | ||
| AK293607.1 | - | 6 | 833 | |||
| AK294039.1 | - | 6 | 815 | |||
| AY033596.1 | - | 5 | 503 | |||
| BC022520.1 | - | 6 | 968 | 4793693 | ||