Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JMJD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114632.1 | NP_001108104.1 | 8 | 1395 | ||
GenBank mRNA | AK000550.1 | - | 8 | 1494 | ||
AK000663.1 | - | 8 | 1369 | |||
AK000814.1 | - | 6 | 1356 | |||
AK225503.1 | - | 8 | 1369 | |||
BC025290.1 | - | 8 | 1357 | |||
BC139899.1 | - | 8 | 1374 |
Gene Symbol: JMJD7-PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209150.1 | - | 7 | 1614 | ||
AF065216.1 | - | 7 | 1446 |