Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPATA31A7 SIRNA Assays ›
Gene Symbol:
SPATA31A7
SPATA31A1
SPATA31A3
SPATA31A5
Gene Name:
SPATA31 subfamily A member 7
SPATA31 subfamily A member 1
SPATA31 subfamily A member 3
SPATA31 subfamily A member 5
Gene Aliases:
AEP1, C9orf36A, FAM75A4, FAM75A7, SPATA31A4
C9orf36, C9orf36A, FAM75A1, FAM75A2, SPATA31A2
FAM75A3
FAM75A5
Chromosome Location:
Chr.9: 61190036 - 61196280 on Build GRCh38
Chromosome Location:
Chr.9: 39355667 - 39361962 on Build GRCh38
Chromosome Location:
Chr.9: 66986301 - 66992550 on Build GRCh38
Chromosome Location:
Chr.9: 60914372 - 60920653 on Build GRCh38
UniGene ID:
Hs.123004
Hs.575677
Hs.632078
Hs.645497
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID
s230502
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPATA31A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015667.2
NP_056482.2
4
3806
GenBank mRNA
BC040528.1
-
4
3806
4838470
Gene Symbol: SPATA31A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085452.2
NP_001078921.2
4
3839
XM_011518006.1
XP_011516308.1
4
3877
GenBank mRNA
AK302569.1
-
4
3839
BC146855.1
-
4
3798
9056783
Gene Symbol: SPATA31A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083124.1
NP_001076593.1
4
3806
XM_011545680.2
XP_011543982.1
3
3723
GenBank mRNA
AL080148.1
-
1
2761
Gene Symbol: SPATA31A5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113541.1
NP_001107013.1
4
3806
XM_011545671.2
XP_011543973.1
4
3877
Back To Top
More Information
Gene Ontology Categories:
Function(s)
spermatogenesis
cell differentiation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System