Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DNAH14 SIRNA Assays ›
Gene Symbol:
DNAH14
Gene Name:
dynein axonemal heavy chain 14
Gene Aliases:
C1orf67, Dnahc14, HL-18, HL18
Chromosome Location:
Chr.1: 224928422 - 225399294 on Build GRCh38
UniGene ID:
Hs.133977
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230526
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DNAH14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145154.1
NP_001138626.1
6
814
NM_001373.1
NP_001364.1
6
814
NM_144989.2
NP_659426.2
6
807
XM_006711735.3
XP_006711798.1
6
2045
XM_011544057.2
XP_011542359.1
6
2045
XM_011544058.2
XP_011542360.1
6
2045
XM_011544059.2
XP_011542361.1
6
2045
XM_011544061.2
XP_011542363.1
6
2045
XM_011544062.2
XP_011542364.1
6
2045
XM_011544063.2
XP_011542365.1
6
2045
XM_011544064.2
XP_011542366.1
6
2045
XM_011544065.2
XP_011542367.1
6
2045
XM_011544066.2
XP_011542368.1
6
2045
XM_011544067.2
XP_011542369.1
6
2045
XM_011544069.2
XP_011542371.1
6
2045
XM_011544070.2
XP_011542372.1
6
2045
XM_011544071.2
XP_011542373.1
6
2046
XM_011544072.2
XP_011542374.1
6
2045
XM_017000295.1
XP_016855784.1
6
2045
XM_017000296.1
XP_016855785.1
6
2045
XM_017000297.1
XP_016855786.1
6
2045
XM_017000298.1
XP_016855787.1
6
2044
XM_017000300.1
XP_016855789.1
6
2039
GenBank mRNA
BC017415.2
-
5
633
BC119716.1
-
6
801
BC119717.1
-
6
801
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
microtubule motor activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bp9xj:80/100.66.77.12:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0