Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATXN3 SIRNA Assays ›
          Gene Symbol:
          ATXN3 ATXN8
          Gene Name:
          ataxin 3
          ataxin 8
          Gene Aliases:
          AT3, ATX3, JOS, MJD, MJD1, SCA3
          ATXN3
          Chromosome Location:
          Chr.14: 92058552 - 92106621 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.532632
          Hs.532632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s230539

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATXN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127696.1 NP_001121168.1 5, 6 492
          NM_001127697.2 NP_001121169.2 4, 5 384
          NM_001164779.1 NP_001158251.1 2, 3 174
          NM_001164780.1 NP_001158252.1 3, 4 241
          NM_001164781.1 NP_001158253.1 4, 5 327
          NM_004993.5 NP_004984.2 6, 7 537
          NM_030660.4 NP_109376.1 5, 6 372
          NR_028453.1 5, 6 451
          NR_028454.1 4, 5 286
          NR_028455.1 5, 6 451
          NR_028456.1 4, 5 286
          NR_028459.1 6, 7 537
          NR_028464.1 4, 5 384
          NR_028465.1 4, 5 406
          NR_028468.1 5, 6 451
          NR_028469.1 4, 5 286
          GenBank mRNA AB050194.1 - 5, 6 361
          AK225884.1 - 6, 7 476
          AK307720.1 - 3, 4 202
          AK314919.1 - 6, 7 489
          BC022245.1 - 5, 6 420
          BC033711.1 - 6, 7 480 4393766
          BC095402.1 - 6, 7 497 30347215
          BU190081.1 - 5, 6 471
          BU663113.1 - 3, 4 291
          GQ176433.1 - 6, 7 490
          GQ176434.1 - 6, 7 489
          GQ176435.1 - 6, 7 490
          GQ176436.1 - 4, 5 337
          GQ176437.1 - 4, 5 337
          GQ176438.1 - 5, 6 404
          GQ176440.1 - 5, 6 325
          GQ176441.1 - 5, 6 404
          GQ176442.1 - 4, 5 239
          GQ176443.1 - 5, 6 404
          GQ176445.1 - 4, 5 239
          GQ176446.1 - 4, 5 236
          GQ176451.1 - 4, 5 239
          GQ176452.1 - 5, 6 490
          GQ176454.1 - 6, 7 490
          GQ176455.1 - 5, 6 325
          GQ176456.1 - 4, 5 239
          GQ176457.1 - 4, 5 337
          GQ176458.1 - 5, 6 404
          GQ176459.1 - 2, 3 127
          GQ176462.1 - 6, 7 490
          GQ176464.1 - 5, 6 404
          GQ176466.1 - 4, 5 239
          GQ176467.1 - 5, 6 325
          GQ176470.1 - 4, 5 359
          GQ176474.1 - 4, 5 337
          GQ176475.1 - 4, 5 239
          GQ176476.1 - 6, 7 490
          GQ176477.1 - 5, 6 404
          GQ176479.1 - 5, 6 404
          GQ176480.1 - 5, 6 404
          GQ176481.1 - 5, 6 404
          GQ176483.1 - 5, 6 404
          GQ176484.1 - 5, 6 404
          GQ176485.1 - 4, 5 337
          GQ176486.1 - 5, 6 445
          GQ176487.1 - 4, 5 337
          GQ176488.1 - 6, 7 490
          GQ176489.1 - 5, 6 445
          GQ176490.1 - 4, 5 371
          GQ176492.1 - 5, 6 404
          GQ176493.1 - 4, 5 336
          GQ176495.1 - 4, 5 337
          GQ176496.1 - 4, 5 239
          GQ176497.1 - 5, 6 404
          GQ176498.1 - 5, 6 325
          GQ176499.1 - 6, 7 490
          GQ176502.1 - 5, 6 404
          GQ176503.1 - 4, 5 238
          GQ176504.1 - 6, 7 490
          GQ176505.1 - 4, 5 239
          GQ176506.1 - 4, 5 239
          GQ176507.1 - 5, 6 403
          GQ176508.1 - 6, 7 490
          GQ176509.1 - 4, 5 239
          GQ176510.1 - 6, 7 490
          GQ176511.1 - 4, 5 337
          GQ176512.1 - 6, 7 487
          GQ176513.1 - 5, 6 404
          GQ176514.1 - 3, 4 194
          GQ176515.1 - 4, 5 238
          GQ176516.1 - 6, 7 490
          GQ176519.1 - 5, 6 403
          GQ176520.1 - 3, 4 194
          GQ176521.1 - 4, 5 239
          GQ176522.1 - 4, 5 239
          GQ176523.1 - 6, 7 490
          GQ176525.1 - 4, 5 239
          GQ176526.1 - 4, 5 280
          GQ176527.1 - 5, 6 404
          GQ176528.1 - 5, 6 404
          GQ176530.1 - 5, 6 404
          GQ176531.1 - 6, 7 490
          GQ176532.1 - 5, 6 490
          GQ176533.1 - 6, 7 490
          GQ176535.1 - 239
          GQ176536.1 - 5, 6 325
          GQ176537.1 - 5, 6 404
          GQ176538.1 - 6, 7 490
          GQ176539.1 - 5, 6 325
          GQ176540.1 - 5, 6 378
          GQ176542.1 - 2, 3 127
          GQ176543.1 - 4, 5 239
          GQ176544.1 - 4, 5 239
          GQ176546.1 - 5, 6 404
          GQ176547.1 - 4, 5 239
          GQ176549.1 - 4, 5 239
          GQ176550.1 - 6, 7 490
          GQ176551.1 - 6, 7 489
          GQ176552.1 - 6, 7 489
          GQ176553.1 - 5, 6 404
          GQ176554.1 - 4, 5 239
          GQ176556.1 - 4, 5 239
          GQ176558.1 - 5, 6 404
          GQ176559.1 - 4, 5 239
          GQ176560.1 - 6, 7 490
          GQ176563.1 - 6, 7 490
          GQ176565.1 - 6, 7 490
          GQ176566.1 - 5, 6 325
          GQ176567.1 - 6, 7 490
          GQ176568.1 - 6, 7 490
          GQ176569.1 - 5, 6 401
          GQ176570.1 - 6, 7 490
          GQ176571.1 - 4, 5 239
          GQ176572.1 - 5, 6 404
          GQ176573.1 - 5, 6 325
          GQ176577.1 - 6, 7 490
          GQ176578.1 - 6, 7 490
          GQ176580.1 - 6, 7 490
          GQ176585.1 - 4, 5 238
          GQ176586.1 - 5, 6 404
          GQ176587.1 - 4, 5 239
          GQ176588.1 - 5, 6 404
          GQ176589.1 - 6, 7 490
          GQ176591.1 - 4, 5 239
          GQ176592.1 - 5, 6 324
          GQ176594.1 - 6, 7 490
          GQ176595.1 - 5, 6 404
          GQ176596.1 - 5, 6 404
          GQ176597.1 - 6, 7 490
          GQ176599.1 - 6, 7 490
          GQ176601.1 - 4, 5 239
          GQ176602.1 - 6, 7 508
          GQ176603.1 - 4, 5 239
          GQ176604.1 - 4, 5 239
          GQ176605.1 - 5, 6 404
          GQ176607.1 - 5, 6 404
          GQ176608.1 - 4, 5 337
          GQ176609.1 - 490
          GQ176610.1 - 5, 6 404
          GQ176611.1 - 5, 6 404
          GQ176612.1 - 6, 7 490
          GQ176614.1 - 3, 4 194
          GQ176615.1 - 490
          GQ176616.1 - 6, 7 490
          GQ176617.1 - 5, 6 404
          GQ176618.1 - 316
          GQ176619.1 - 6, 7 490
          GQ176620.1 - 5, 6 404
          GQ176621.1 - 6, 7 490
          GQ176622.1 - 6, 7 490
          GQ176623.1 - 4, 5 239
          GQ176624.1 - 5, 6 423
          GQ176626.1 - 4, 5 239
          GQ176627.1 - 2, 3 127
          GQ176628.1 - 490
          GQ176629.1 - 404
          GQ176630.1 - 5, 6 404
          GQ176631.1 - 4, 5 337
          GQ176632.1 - 6, 7 490
          GQ176633.1 - 2, 3 127
          GQ176635.1 - 239
          GQ176636.1 - 3, 4 311
          GQ176637.1 - 4, 5 239
          GQ176638.1 - 4, 5 239
          GQ176639.1 - 4, 5 337
          GQ176640.1 - 6, 7 490
          GQ176641.1 - 210
          GQ176642.1 - 6, 7 490
          GQ176643.1 - 239
          GQ176644.1 - 6, 7 485
          GQ176645.1 - 6, 7 490
          GQ176646.1 - 6, 7 490
          GQ176647.1 - 239
          GQ176648.1 - 5, 6 404
          GQ176649.1 - 5, 6 404
          GQ176653.1 - 3, 4 194
          GQ176655.1 - 4, 5 334
          GQ176656.1 - 4, 5 332
          GQ176658.1 - 6, 7 490
          GQ176660.1 - 5, 6 404
          GQ176661.1 - 3, 4 194
          GQ176662.1 - 5, 6 404
          GQ176663.1 - 4, 5 337
          GQ176664.1 - 5, 6 404
          GQ176665.1 - 5, 6 325
          GQ176666.1 - 3, 4 194
          GQ176668.1 - 6, 7 487
          GQ176669.1 - 404
          GQ176670.1 - 3, 4 292
          GQ176671.1 - 490
          GQ176673.1 - 6, 7 490
          GQ176674.1 - 6, 7 490
          GQ176675.1 - 5, 6 325
          GQ176676.1 - 4, 5 239
          GQ176677.1 - 6, 7 490
          GQ176678.1 - 5, 6 404
          GQ176679.1 - 5, 6 325
          GQ176680.1 - 6, 7 490
          GQ176682.1 - 5, 6 404
          GQ176684.1 - 6, 7 490
          GQ176685.1 - 6, 7 489
          GQ176686.1 - 490
          GQ176687.1 - 6, 7 490
          GQ176688.1 - 4, 5 337
          GQ176689.1 - 3, 4 194
          GQ176690.1 - 5, 6 404
          GQ176692.1 - 6, 7 490
          GQ176693.1 - 490
          GQ176694.1 - 5, 6 325
          GQ176695.1 - 4, 5 337
          GQ176696.1 - 5, 6 404
          GQ176697.1 - 6, 7 490
          GQ176698.1 - 490
          GQ176699.1 - 490
          GQ176700.1 - 5, 6 325
          GQ176701.1 - 6, 7 490
          GQ176702.1 - 3, 4 194
          GQ176703.1 - 6, 7 490
          GQ176704.1 - 6, 7 490
          GQ176705.1 - 2, 3 127
          GQ176706.1 - 5, 6 404
          GQ176707.1 - 236
          GQ176709.1 - 401
          GQ176710.1 - 2, 3 127
          GQ176711.1 - 3, 4 194
          GQ176713.1 - 239
          GQ176714.1 - 6, 7 490
          GQ176716.1 - 4, 5 239
          GQ176717.1 - 404
          GQ176719.1 - 490
          GQ176720.1 - 4, 5 239
          GQ176721.1 - 6, 7 490
          GQ176722.1 - 5, 6 404
          GQ176723.1 - 3, 4 194
          GQ176724.1 - 4, 5 337
          GQ176725.1 - 6, 7 490
          GQ176726.1 - 4, 5 239
          GQ176727.1 - 2, 3 127
          GQ176728.1 - 6, 7 490
          GQ176729.1 - 4, 5 239
          GQ176731.1 - 4, 5 239
          GQ176732.1 - 239
          GQ176733.1 - 5, 6 404
          GQ176734.1 - 337
          GQ176735.1 - 6, 7 490
          GQ176736.1 - 239
          GQ176737.1 - 6, 7 490
          GQ176738.1 - 5, 6 404
          GQ176739.1 - 404
          GQ176740.1 - 4, 5 239
          GQ176741.1 - 490
          GQ176742.1 - 172
          GQ176743.1 - 4, 5 239
          GQ176744.1 - 5, 6 445
          GQ176745.1 - 490
          GQ176746.1 - 6, 7 490
          GQ176747.1 - 490
          GQ176748.1 - 4, 5 239
          GQ176749.1 - 6, 7 490
          GQ176750.1 - 5, 6 325
          GQ176751.1 - 239
          GQ176753.1 - 3, 4 194
          GQ176754.1 - 5, 6 325
          GQ176755.1 - 5, 6 404
          GQ176756.1 - 6, 7 490
          GQ176757.1 - 4, 5 337
          GQ176758.1 - 4, 5 239
          GQ176760.1 - 5, 6 404
          GQ176761.1 - 3, 4 172
          GQ176762.1 - 490
          GQ176763.1 - 6, 7 490
          GQ176764.1 - 239
          GQ176765.1 - 4, 5 239
          GQ176766.1 - 6, 7 490
          GQ176767.1 - 5, 6 404
          GQ176768.1 - 4, 5 239
          GQ176769.1 - 3, 4 292
          GQ176770.1 - 4, 5 239
          GQ176771.1 - 6, 7 490
          GQ176772.1 - 4, 5 337
          GQ176773.1 - 4, 5 337
          GQ176774.1 - 4, 5 239
          GQ176776.1 - 490
          GQ176778.1 - 3, 4 194
          GQ176779.1 - 4, 5 337
          GQ176780.1 - 4, 5 239
          GQ176781.1 - 4, 5 337
          GQ176782.1 - 4, 5 337
          GQ176783.1 - 239
          GQ176784.1 - 4, 5 337
          GQ176785.1 - 3, 4 172
          GQ176786.1 - 4, 5 337
          GQ176787.1 - 6, 7 490
          GQ176788.1 - 4, 5 239
          GQ176790.1 - 4, 5 336
          GQ176791.1 - 6, 7 490
          GQ176792.1 - 4, 5 334
          GQ176793.1 - 4, 5 337
          GQ176794.1 - 194
          GQ176795.1 - 4, 5 239
          GQ176796.1 - 404
          GQ176797.1 - 5, 6 404
          GQ176798.1 - 404
          GQ176799.1 - 172
          GQ176800.1 - 490
          GQ176801.1 - 404
          GQ176802.1 - 337
          GQ176803.1 - 404
          GQ176804.1 - 127
          GQ176805.1 - 6, 7 466
          GQ176807.1 - 5, 6 404
          GQ176808.1 - 490
          GQ176809.1 - 3, 4 292
          GQ176810.1 - 5, 6 402
          GQ176812.1 - 4, 5 337
          GQ176813.1 - 5, 6 325
          GQ176815.1 - 4, 5 338
          GQ176816.1 - 404
          GQ176819.1 - 5, 6 404
          GQ176820.1 - 6, 7 490
          GQ176821.1 - 4, 5 238
          GQ176822.1 - 5, 6 325
          GQ176824.1 - 6, 7 490
          GQ176827.1 - 6, 7 490
          GQ176828.1 - 4, 5 239
          GQ176829.1 - 6, 7 490
          GQ176830.1 - 461
          GQ176831.1 - 490
          GQ176832.1 - 4, 5 239
          GQ176833.1 - 5, 6 404
          GQ176834.1 - 403
          GQ176835.1 - 5, 6 324
          GQ176836.1 - 5, 6 325
          GQ176837.1 - 6, 7 490
          GQ176838.1 - 6, 7 490
          GQ176839.1 - 239
          GQ176840.1 - 4, 5 239
          GQ176841.1 - 6, 7 490
          GQ176842.1 - 404
          GQ176843.1 - 4, 5 238
          GQ176844.1 - 4, 5 252
          GQ176845.1 - 5, 6 403
          GQ176846.1 - 6, 7 490
          GQ176847.1 - 4, 5 236
          S75313.1 - 6, 7 503
          U64820.1 - 6, 7 525
          U64821.1 - 6, 7 488
          U64822.1 - 5, 6 465
          Gene Symbol: ATXN8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA GQ176777.1 - 172

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          nucleotide-excision repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          ubiquitin-dependent protein catabolic process
          misfolded or incompletely synthesized protein catabolic process
          chemical synaptic transmission
          nervous system development
          regulation of cell-substrate adhesion
          protein deubiquitination
          actin cytoskeleton organization
          cellular response to heat
          monoubiquitinated protein deubiquitination
          exploration behavior
          proteasome-mediated ubiquitin-dependent protein catabolic process
          intermediate filament cytoskeleton organization
          protein K63-linked deubiquitination
          histone H3 deacetylation
          protein K48-linked deubiquitination
          cellular response to misfolded protein
          positive regulation of ERAD pathway
          protein localization to cytosolic proteasome complex involved in ERAD pathway
          RNA polymerase II regulatory region DNA binding
          histone deacetylase activity
          thiol-dependent ubiquitin-specific protease activity
          protein binding
          omega peptidase activity
          ubiquitin protein ligase binding
          identical protein binding
          ATPase binding
          Lys63-specific deubiquitinase activity
          Lys48-specific deubiquitinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cvnbf:80/100.66.76.207:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0