Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYO1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101421.3 | NP_001094891.3 | 11 | 1224 | ||
XM_011538223.2 | XP_011536525.1 | 11 | 1666 | |||
XM_017019207.1 | XP_016874696.1 | 11 | 1666 | |||
GenBank mRNA | DV080459.1 | - | 3 | 164 |