Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TP73-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033710.1 | 4 | 2220 | |||
NR_033711.1 | 4 | 2220 | ||||
GenBank mRNA | AB007964.1 | - | 4 | 2170 | ||
AL133648.1 | - | 4 | 1473 |