Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TACC1 SIRNA Assays ›
          Gene Symbol:
          TACC1
          Gene Name:
          transforming acidic coiled-coil containing protein 1
          Gene Aliases:
          Ga55
          Chromosome Location:
          Chr.8: 38728186 - 38853028 on Build GRCh38
          UniGene ID:
          Hs.279245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s230694

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TACC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122824.1 NP_001116296.1 6, 7 1073
          NM_001146216.2 NP_001139688.1 8, 9 2249
          NM_006283.2 NP_006274.2 8, 9 2303
          XM_005273625.4 XP_005273682.1 10, 11 2453
          XM_005273628.1 XP_005273685.1 9, 10 1960
          XM_005273629.1 XP_005273686.1 10, 11 1973
          XM_005273630.1 XP_005273687.1 9, 10 1981
          XM_011544631.1 XP_011542933.1 9, 10 2339
          XM_011544632.2 XP_011542934.1 11, 12 2514
          XM_011544633.1 XP_011542935.1 9, 10 2252
          XM_011544634.1 XP_011542936.1 8, 9 2216
          XM_011544635.1 XP_011542937.1 9, 10 2043
          XM_011544636.2 XP_011542938.1 6, 7 699
          XM_011544637.1 XP_011542939.1 6, 7 986
          XM_011544638.2 XP_011542940.1 6, 7 589
          XM_017013766.1 XP_016869255.1 8, 9 1924
          XM_017013767.1 XP_016869256.1 9, 10 1937
          XM_017013768.1 XP_016869257.1 8, 9 1837
          XM_017013769.1 XP_016869258.1 9, 10 1850
          XM_017013770.1 XP_016869259.1 8, 9 1873
          XM_017013771.1 XP_016869260.1 8, 9 2145
          XM_017013772.1 XP_016869261.1 7, 8 1837
          XM_017013773.1 XP_016869262.1 7, 8 2104
          XM_017013774.1 XP_016869263.1 7, 8 1789
          XM_017013775.1 XP_016869264.1 8, 9 1858
          XM_017013776.1 XP_016869265.1 9, 10 1850
          XM_017013777.1 XP_016869266.1 7, 8 2014
          XM_017013778.1 XP_016869267.1 7, 8 1713
          GenBank mRNA AB029026.1 - 7, 8 873
          AF049910.1 - 8, 9 2244
          AK294931.1 - 8, 9 1817
          AK295071.1 - 8, 9 1851
          AK304507.1 - 10, 11 2049
          AK304725.1 - 5, 6 561
          AK304774.1 - 6, 7 623
          AK308849.1 - 6, 7 557
          AK314620.1 - 8, 9 2303
          AY039239.1 - 7, 8 613
          AY177411.1 - 6, 7 1014
          BC041391.1 - 10, 11 2413 5276767
          BX647696.1 - 8, 9 2248

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          cell cycle
          cell proliferation
          cerebral cortex development
          cell division
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0