Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230695
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122824.1
NP_001116296.1
8
1248
NM_001146216.2
NP_001139688.1
10
2424
NM_006283.2
NP_006274.2
10
2478
XM_005273625.4
XP_005273682.1
12
2628
XM_005273628.1
XP_005273685.1
11
2135
XM_005273629.1
XP_005273686.1
12
2148
XM_005273630.1
XP_005273687.1
11
2156
XM_011544631.1
XP_011542933.1
11
2514
XM_011544632.2
XP_011542934.1
13
2689
XM_011544633.1
XP_011542935.1
11
2427
XM_011544634.1
XP_011542936.1
10
2391
XM_011544635.1
XP_011542937.1
11
2218
XM_011544636.2
XP_011542938.1
8
874
XM_011544637.1
XP_011542939.1
8
1161
XM_011544638.2
XP_011542940.1
8
764
XM_017013766.1
XP_016869255.1
10
2099
XM_017013767.1
XP_016869256.1
11
2112
XM_017013768.1
XP_016869257.1
10
2012
XM_017013769.1
XP_016869258.1
11
2025
XM_017013770.1
XP_016869259.1
10
2048
XM_017013771.1
XP_016869260.1
10
2320
XM_017013772.1
XP_016869261.1
9
2012
XM_017013773.1
XP_016869262.1
9
2279
XM_017013774.1
XP_016869263.1
9
1964
XM_017013775.1
XP_016869264.1
10
2033
XM_017013776.1
XP_016869265.1
11
2025
XM_017013777.1
XP_016869266.1
9
2189
XM_017013778.1
XP_016869267.1
9
1888
GenBank mRNA
AB029026.1
-
9
1045
AF049910.1
-
10
2419
AK294931.1
-
10
1992
AK295071.1
-
10
2026
AK304507.1
-
12
2224
AK304725.1
-
7
733
AK304774.1
-
8
798
AK314620.1
-
10
2478
AY039239.1
-
9
788
AY139007.1
-
2
1123
AY177411.1
-
8
1189
BC041391.1
-
12
2588
5276767
BX647696.1
-
10
2423
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0