Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GTF2H2C SIRNA Assays ›
          Gene Symbol:
          GTF2H2C GTF2H2 GTF2H2B GTF2H2C_2
          Gene Name:
          GTF2H2 family member C
          general transcription factor IIH subunit 2
          general transcription factor IIH subunit 2B (pseudogene)
          GTF2H2 family member C, copy 2
          Gene Aliases:
          GTF2H2C_2
          BTF2, BTF2P44, T-BTF2P44, TFIIH, p44
          -
          GTF2H2D
          Chromosome Location:
          Chr.5: 69560136 - 69592903 on Build GRCh38
          Chromosome Location:
          Chr.5: 71035124 - 71109278 on Build GRCh38
          Chromosome Location:
          Chr.5: 70415370 - 70450362 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.191356
          Hs.191356
          Hs.607501
          Hs.422901
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s230758

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GTF2H2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098728.1 NP_001092198.1 5 424
          XM_005248588.4 XP_005248645.1 5 351
          XM_005248590.3 XP_005248647.1 5 289
          XM_005248592.3 XP_005248649.1 3 153
          XM_006714688.3 XP_006714751.1 5 342
          XM_011543615.1 XP_011541917.1 5 424
          XM_017009804.1 XP_016865293.1 3 207
          GenBank mRNA AK292361.1 - 5 403
          AK315282.1 - 5 424
          BC105980.1 - 5 282 4445643
          BC157894.1 - 5 354
          BC171860.1 - 6 416 9054514
          Gene Symbol: GTF2H2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001515.3 NP_001506.1 5 421
          XM_017009403.1 XP_016864892.1 6 1030
          XM_017009404.1 XP_016864893.1 6 518
          XM_017009405.1 XP_016864894.1 6 373
          XM_017009406.1 XP_016864895.1 5 276
          GenBank mRNA AF078847.1 - 5 406
          AK296578.1 - 5 425
          BC005345.1 - 5 279
          BM083743.1 - 5 421
          BT006773.1 - 4 211
          Z30094.1 - 4 211
          Gene Symbol: GTF2H2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033417.1 4 388
          GenBank mRNA BC171919.1 - 4 317 9054573
          BX537982.1 - 4 228
          Gene Symbol: GTF2H2C_2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042490.3 NP_001035955.1 6 499
          GenBank mRNA BC064557.1 - 5 379 5549265
          BC065021.1 - 6 500 6047020
          BX647445.1 - 5 264

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nucleotide-excision repair, DNA duplex unwinding
          G-protein coupled receptor internalization
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          translation
          protein phosphorylation
          response to UV
          nucleotide-excision repair, DNA incision
          global genome nucleotide-excision repair
          protein binding
          zinc ion binding
          nucleic acid binding
          transcription factor activity, sequence-specific DNA binding
          protein kinase activity
          DNA-dependent ATPase activity
          translation factor activity, RNA binding
          RNA polymerase II carboxy-terminal domain kinase activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0