Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 8 | 1085 | ||
NM_022552.4 | NP_072046.2 | 13 | 1771 | |||
NM_153759.3 | NP_715640.2 | 9 | 1055 | |||
NM_175629.2 | NP_783328.1 | 13 | 1842 | |||
NR_135490.1 | 13 | 1842 | ||||
XM_005264175.4 | XP_005264232.1 | 13 | 1705 | |||
XM_005264177.4 | XP_005264234.1 | 8 | 997 | |||
XM_011532662.2 | XP_011530964.1 | 11 | 1707 | |||
XM_011532663.2 | XP_011530965.1 | 14 | 1884 | |||
XM_011532664.2 | XP_011530966.1 | 13 | 1782 | |||
XM_011532666.2 | XP_011530968.1 | 9 | 1070 | |||
XM_011532667.2 | XP_011530969.1 | 8 | 887 | |||
XM_017003526.1 | XP_016859015.1 | 13 | 1813 | |||
XM_017003527.1 | XP_016859016.1 | 8 | 1041 | |||
GenBank mRNA | AB208833.1 | - | 13 | 1741 | ||
AF067972.2 | - | 13 | 1733 | |||
AF331856.1 | - | 13 | 1721 | |||
AF480163.1 | - | 9 | 1051 | |||
AK025230.1 | - | 3 | 95 | |||
BC043617.1 | - | 13 | 1741 | 6150112 |