Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KALRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322989.1 | NP_001309918.1 | 34 | 5937 | ||
NM_001322990.1 | NP_001309919.1 | 34 | 5925 | |||
NM_001322991.1 | NP_001309920.1 | 34 | 5898 | |||
NM_001322992.1 | NP_001309921.1 | 33 | 5814 | |||
NM_003947.5 | NP_003938.1 | 34 | 5964 | |||
XM_006713815.3 | XP_006713878.1 | 34 | 6212 | |||
XM_017007433.1 | XP_016862922.1 | 34 | 6185 | |||
XM_017007434.1 | XP_016862923.1 | 34 | 6173 | |||
XM_017007435.1 | XP_016862924.1 | 34 | 6146 | |||
GenBank mRNA | AK126954.1 | - | 1 | 2853 | ||
U94190.1 | - | 34 | 5907 |